Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(3-aminophenoxy)-5-chlorobenzene Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. B35505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35505G 5 g
B35501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35505G Supplier TCI America Supplier No. B35505G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99503-73-8
Molecular Formula C18H17Cl3N2O2
Molecular Weight (g/mol) 399.70
MDL Number MFCD14708170
InChI Key YZUFDVOYHZOIGU-UHFFFAOYSA-N
PubChem CID 44630035
IUPAC Name 3-[3-(3-aminophenoxy)-5-chlorophenoxy]aniline dihydrochloride
SMILES Cl.Cl.NC1=CC(OC2=CC(OC3=CC=CC(N)=C3)=CC(Cl)=C2)=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 399.70
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(3-aminophenoxy)-5-chlorobenzene Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.