CAS RN 99534-03-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ PKA Inhibitor 5-24
The PKA Inhibitor 5-24, also referenced under CAS 99534-03-9, controls the biological activity of PKA.
2
Tocris Bioscience™ PKI (5-24)
High affinity PKA inhibitor