Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PKA Inhibitor 5-24

Catalog No. 116805500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-680-5500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-680-5500UG Supplier MilliporeSigma™ Supplier No. 116805500UG
Add to Cart
Add to Cart

The PKA Inhibitor 5-24, also referenced under CAS 99534-03-9, controls the biological activity of PKA.

Peptide Sequence: H-Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp-OH
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt. Sold on the basis of peptide content.
Description 95% by HPLC
Target Proteins PKA
CAS 99534-03-9
Molecular Formula C94H148N32O31
Formula Weight 2222.4g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.