accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CNH3CCH3NCH3H3CNH3CCH3

CAS RN 996-70-3

IUPAC Name: dimethyl[1,2,2-tris(dimethylamino)ethenyl]amine
Synonyms: dimethyl[1,2,2-tris(dimethylamino)ethenyl]amine
Molecular Weight (g/mol): 200.33
Molecular Formula: C10H24N4
InChi Key: CBXRMKZFYQISIV-UHFFFAOYSA-N
SMILES: CN(C)C(N(C)C)=C(N(C)C)N(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)