Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrakis(Dimethylamino)Ethylene

Catalog No. 67461350G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-461-350G 50 g
67-461-31G 1 g
67-461-310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 67-461-350G Supplier Sigma Aldrich Supplier No. 67461350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used to convert +a-bromoketones into 1,4-diketones.

Specifications

CAS 996-70-3
Density 0.861 g/mL at 25°C
Boiling Point 59°C (0.9 mmHg, lit.)
Molecular Formula C10H24N4
Refractive Index n20/D 1.48
Linear Formula [(CH3)2N]2C=C[N(CH3)2]2
MDL Number MFCD00009601
Quantity 50 g
Synonym Octamethylethylenetetramine; TDAE
Molecular Weight (g/mol) 200.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.