accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 996-98-5

H2NHNOONHNH2

CAS RN 996-98-5

IUPAC Name: ethanedihydrazide
Synonyms: oxalyldihydrazide
Molecular Weight (g/mol): 118.1
Molecular Formula: C2H6N4O2
InChi Key: SWRGUMCEJHQWEE-UHFFFAOYSA-N
SMILES: NNC(=O)C(=O)NN
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results