Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalyl Dihydrazide 96.0+%, TCI America™
SDP

Catalog No. O008325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O008325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O008325G Supplier TCI America Supplier No. O008325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 996-98-5
Melting Point 244°C
Color White-Yellow
Molecular Formula C2H6N4O2
Quantity 25 g
MDL Number MFCD00007608
Synonym oxalyl dihydrazide, oxalic dihydrazide, oxaldihydrazide, ethanedioic acid, dihydrazide, oxalyldihydrazide, oxalhydrazide, oxalylhydrazine, oxaloylhydrazine, oxalic hydrazide, oxalyl hydrazide
InChI Key SWRGUMCEJHQWEE-UHFFFAOYSA-N
SMILES NNC(=O)C(=O)NN
IUPAC Name ethanedihydrazide
Molecular Weight (g/mol) 118.10
PubChem CID 13826
Formula Weight 118.10
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Oxalyl Dihydrazide
Show More Show Less
EINECSNumber (2)-0923
RTECSNumber RO2840000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.