accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99627-05-1

IUPAC Name: 3,4,5-trifluorophenol
Synonyms: 3,4,5-trifluorophenol
Molecular Weight (g/mol): 148.08
Molecular Formula: C6H3F3O
InChi Key: ZRTWIJKGTUGZJY-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results