Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trifluorophenol 98.0+%, TCI America™
SDP

Catalog No. T22995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22995G 5 g
T229925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22995G Supplier TCI America Supplier No. T22995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99627-05-1
Molecular Formula C6H3F3O
Molecular Weight (g/mol) 148.084
MDL Number MFCD00042427
InChI Key ZRTWIJKGTUGZJY-UHFFFAOYSA-N
PubChem CID 2777943
IUPAC Name 3,4,5-trifluorophenol
SMILES C1=C(C=C(C(=C1F)F)F)O

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 177°C
Quantity 5 g
UN Number 3077
Synonym phenol, 3,4,5-trifluoro, 3,4,5-trifluoro-phenol, pubchem2316, 3,4,5-trifluoropheno, acmc-209sdi, ksc486m6r, 3 , 4 , 5-trifluorophenol, phenol, 3,4,5-trifluoro-9ci, 3,4,5-trifluorophenol
Formula Weight 148.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trifluorophenol
EINECSNumber (3)-4667
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.