CAS RN 99755-59-6
CAS RN 99755-59-6
IUPAC Name:
6-{propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol
Synonyms:
rotigotine transdermal patch rotigotine
Molecular Weight (g/mol):
315.48
Molecular Formula:
C19H25NOS
InChi Key:
KFQYTPMOWPVWEJ-UHFFFAOYNA-N
SMILES:
CCCN(CCC1=CC=CS1)C1CCC2=C(O)C=CC=C2C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Rotigotine
CAS: 99755-59-6 Molecular Formula: C19H25NOS