accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOONH3C

CAS RN 99762-80-8

IUPAC Name: 5,12-dibutyl-5,7,12,14-tetrahydro-5,12-diazapentacene-7,14-dione
Synonyms: 5,12-dibutyl-5,12-diazapentacene-7,14-dione
Molecular Weight (g/mol): 424.54
Molecular Formula: C28H28N2O2
InChi Key: MYFHGRHNRNCMHV-UHFFFAOYSA-N
SMILES: CCCCN1C2=CC=CC=C2C(=O)C2=CC3=C(C=C12)C(=O)C1=CC=CC=C1N3CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results