accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99769-29-6

IUPAC Name: ethyl 2-bromobenzene-1-sulfonate
Synonyms: ethyl 2-bromobenzenesulfonate
Molecular Weight (g/mol): 265.12
Molecular Formula: C8H9BrO3S
InChi Key: QUZFLGXJHUNSEH-UHFFFAOYSA-N
SMILES: CCOS(=O)(=O)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results