CAS RN 99953-00-1
CAS RN 99953-00-1
IUPAC Name:
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxy-2,6-dimethylphenyl)propanoic acid
Synonyms:
(2S)-2-[(tert-butoxycarbonyl)amino]-3-(4-hydroxy-2,6-dimethylphenyl)propanoic acid
Molecular Weight (g/mol):
309.36
Molecular Formula:
C16H23NO5
InChi Key:
QSKQZXRPUXGSLR-ZDUSSCGKSA-N
SMILES:
CC1=CC(O)=CC(C)=C1C[C@H](NC(=O)OC(C)(C)C)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-L- 2 6-di-Me Tyr-OH, Advanced ChemTech
CAS: 99953-00-1 Molecular Formula: C16H23NO5 Molecular Weight (g/mol): 309.36 MDL Number: MFCD01860643 InChI Key: QSKQZXRPUXGSLR-ZDUSSCGKSA-N IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxy-2,6-dimethylphenyl)propanoic acid SMILES: CC1=CC(O)=CC(C)=C1C[C@H](NC(=O)OC(C)(C)C)C(O)=O