- TaqMan Array Card: Respiratory Tract Microbiota Comprehensive Card
- OpenArray: Respiratory Tract Microbiota Plate
- TaqMan PreAmp Pool, Respiratory Tract Microbiota
Respiratory Tract Microbiota Detection
Detection of exposure of viral, bacterial, and fungal microbes in the upper and lower respiratory tract is crucial because if undetected and, they could cause serious diseases. Traditional methods of microbiota detection include immunoassays, which have limitations, and culture-based methods that are easily contaminated and often overlook the presence of bacteria and fungi. Molecular testing is an alternative option, but it is expensive and lacks flexibility. Also, these methods focus primarily on detection of either viruses or bacteria, but not both.
With Applied Biosystems™ OpenArray™ and TaqMan® testing solutions, you get superior microbiota detection that includes coverage of 42 verified pathogen targets or customizable options. When processed through Applied Biosystems™ QuantStudio™ Real-Time PCR Systems, the sensitive, flexible-content, syndromic panels enable reliable results. OpenArray and TaqMan Array Card technology is also cost effective and works with multiple sample types.
Contact your Fisher Healthcare product manager to learn more.
Test Features
- Unparalleled Coverage: With 42 verified pathogen targets, Applied Biosystems panels feature the most pathogenic markers of any test on the market.
- Flexible: Verified for TaqMan Array Cards and OpenArray plates and offer customized solutions for 96- and 384-well plates or single-tube assays.
- Affordable: Up to three times more cost effective than other commercially available panel-based molecular respiratory tests.
- Versatile: Accepts multiple sample types, including nasopharyngeal swabs, bronchoalveolar lavage (BAL), and nasopharyngeal aspirate.
TaqMan Assays Included
Controls Included
Data and Support
- TaqMan Respiratory Tract Microbiota Amplification Control
- TaqMan Universal RNA Spike-In/Reverse Transcription (Xeno) Control
- TaqMan Universal Extraction Control Organism
- Application Note: Detecting Respiratory Tract Microbiota Using TaqMan Array Cards
TaqMan Array Card Format
OpenArray Plate Format
| Number of Wells | 384 |
| Number of Targets | 42 Respiratory Plus 4 Control (Xeno, RNase P, and Bacillus atropheus) |
| Number of Samples | 8 |
| Master Mix Volume | 400 μL Per TaqMan Array Card (384 Reactions) |
| Reaction Volume | 1 μL |
| Samples/Day | Up to 64 |
| Compatible Instruments | QuantStudio 12K Flex Real-Time PCR System with TaqMan Array Card Block |
| Time to Results | 5 hours |
| Number of Wells | 2,688 |
| Number of Targets | 31 Respiratory Plus 4 Control (Xeno, RNase P, and Bacillus atropheus) |
| Number of Samples | Up to 24 Samples Per Plate (Spotted in Triplicate) |
| Master Mix Volume | 120 μL Per OpenArray Plate (3,072 Reactions) |
| Reaction Volume | 33 nL |
| Samples/Day | Up to 288 |
| Compatible Instruments | QuantStudio 12K Flex Real-Time PCR System with OpenArray Block |
| Time to Results | 5 hours |
Microbiota Targets
Included with TaqMan Array Card. Those included with OpenArray Plate are marked with an asterisk (*).
| Pathogen Type | Target Pathogen |
|---|---|
| Bacterial | Bordetella (PAN)* |
| Citrobacter freundii | |
| Bordetella holmesii | |
| Bordetella pertussis* | |
| Chlamydophila pneumoniae* | |
| Mycoplasma pneumoniae* | |
| Streptococcus pneumoniae* | |
| Coxiella burnetii | |
| Staphylococcus aureus | |
| Klebsiella pneumoniae* | |
| Legionella pneumophila* | |
| Moraxella catarrhalis | |
| Haemophilus influenzae* | |
| Fungal | Pneumocystis jirovecii |
| Pathogen Type | Target Pathogen |
|---|---|
| Viral | Adenovirus* |
| Coronavirus HKU1* | |
| Coronavirus NL63* | |
| Coronavirus 229E* | |
| Coronavirus OC43* | |
| Human Metapneumovirus* | |
| Rhinovirus* | |
| Enterovirus* | |
| Enterovirus D68 | |
| Influenza A (Pan)* | |
| Influenza A/H1-2009* | |
| Influenza A/H3* | |
| Influenza B* | |
| Parainfluenza 1* | |
| Parainfluenza 2* | |
| Parainfluenza 3* | |
| Parainfluenza 4* | |
| Respiratory Syncytial Virus A* | |
| Respiratory Syncytial Virus B* | |
| Bocavirus* | |
| Epstein-Barr Virus (EBV)* | |
| SARS-CoV | |
| MERS-CoV | |
| Mumps | |
| Measles | |
| Cytomegalovirus* | |
| Human Herpesvirus 6 (HHV-6)* | |
| Varicella Zoster Virus (VZV)* | |
| Parechovirus |
For Research Use Only. Not for use in diagnostic procedures.