UserName
Viewing profile as user:
Techcyte Digital Pathology & AI Product Selection Guide
The United Anatomic and Clinical Pathology AI and Workflow Platform
Streamline your digital pathology workflows for faster, more efficient results.
Access to Techcyte’s end-to-end digital pathology solutions combines scanner technology, image management systems (IMS), and artificial intelligence to enhance diagnostic accuracy and streamline workflows. With Techcyte, your lab can:
  • Scan slides in high resolution for precise image capture
  • Centralize and manage digital slides across anatomic pathology (AP) and clinical pathology (CP) workflows
  • Automate cell identification, classification, and quantification using AI
Techcyte Digital Pathology Workflow
Step 1: Create Slides
Technologists prepare slides following standard preparation protocols, including coverslipping.
Create Slides
Step 2: Scan Slides
Slides are digitized by a scanner, then the whole slide image is uploaded to the Techcyte Fusion Platform image management system.
Scan Slides
Step 3: AI Analysis
Fusion uses both the Techcyte proprietary AI and third-party AI tools to analyze the whole slide image, identifying diagnostically significant areas or objects.
AI Analysis
Step 4: Case Review
A pathologist or laboratory professional reviews the findings and signs out the case.
Case Review
Three-Step Guide to Exploring Digital Pathology and AI Solutions
1. Choose Your Scanner
Select a compatible scanner to digitize slides for high-resolution analysis.
2. Choose an IMS Provider
Techcyte goes beyond traditional image management by supporting and simplifying the entire diagnostic process.
3. Choose AI Software
Techcyte AI streamlines cell detection, classification, and quantification.
ApplicationsScannersIMSWorkflow & AI Solutions
Hematology
Low-Med Throughput:
Pramana,
Grundium, 3DH,
Motic
Med-High Throughput:
Pramana, 3DH,
Hamamatsu
Techcyte Fusion
Techcyte:
  • Blood Differential
  • Digital HemeFISH
  • Bone Marrow Aspirate
  • Flow Cytometry
Microbiology
Low-Med Throughput:
Pramana
Med-High Throughput:
Pramana,
Hamamatsu
Techcyte Fusion
Techcyte:
  • Gram Stains
  • AFB
  • Mycology
  • Body Fluids
ParasitologyLow-Med Throughput:
Pramana,
Grundium
Med-High Throughput:
Pramana,
Hamamatsu, 3DH
Techcyte Fusion
Techcyte:
  • Wet Mount
  • Trichrome
  • Modified Acid Fast
  • Blood Parasites
CytologyLow-Med Throughput:
Pramana,
Grundium
Med-High Throughput:
Pramana,
Hamamatsu, 3DH
Techcyte Fusion
Techcyte:
  • Cervical Cytology
  • Non-GYN
  • Cytology/FNA
HistologyLow-Med Throughput:
Pramana,
Grundium, Motic
Med-High Throughput:
Pramana, 3DH,
Hamamatsu
Techctye Fusion
3DH SlideCenter
(Viewing/Storage)
Techcyte:
  • Frozen Section
  • Organ Donation
  • Dermatology
  • Breast
  • Prostate
  • GI
  • Kidney
  • Live
  • Other Solid Tumors
  • 3rd PartyAI
3DH
  • ICH Quantification
Digital Pathology Slide Scanners
Browse a wide selection of digital pathology slide scanners, which enable faster, high-resolution imaging and efficient sharing of pathology slides, improving diagnostic accuracy, workflow efficiency, and collaboration in clinical and research settings.
Capture High-Resolution Images
See how Epredia solutions use confocal microscopy and digital slide scanning to deliver high-resolution, fluorescence, and 3D imaging for advanced applications.
Epredia
Enable Effortless Scanning
View compact, precise, and fast Grundium digital pathology scanners, designed to make slide scanning effortless, whether for networked clinics or individual labs.
Grundium
Deliver Faster Results
Explore a complete selection of Techcyte digital pathology products, which combine scanner and image management technology with AI to boost productivity.
Techyte
Early Evaluation Program
With Techcyte’s early evaluation, you will get access to the Techcyte platform to process your samples.
Here’s how it works:
  1. Schedule a slide prep meeting with Techcyte to help ensure your slides are optimized for being scanned
  2. Prepare and coverslip your slides, making sure to label them and remove any identifiable information
  3. Send the slides from your institution to the Techcyte lab
  4. Techcyte will scan them and upload them to the platform
Personalized Demo
After shipping your slides, contact Techcyte to schedule your personalized demo meeting to go over how the platform works and review your slides and the associated AI findings. You will be able to compare the manual and digital results and explore Techcyte’s features and benefits.
Fill out the form to get started.
Reach Out to a Representative
Contact a Fisher Healthcare representative to set up a demo of the Unified Anatomic and Clinical Pathology AI and Workflow Platform.
Required Field *
Company
Address
Country
State
Explore More
Grundium Ocus Digital Pathology Case Study
This case study shows how the Grundium Ocus™ Slide Scanner enabled a rural hospital to maintain diagnostic continuity despite a severe pathologist shortage.
Grundium Case Study
How AI and Digital Pathology Are Shaping the Future of Diagnostics
Learn more about the benefits and challenges of implementing AI-powered digital pathology in clinical and research workflows and examine real-world examples of its use.
How AI and Digital Pathology are Shaping the Future of Diagnostics
Webinar: Using Telepathology to Advance Patient Care and Lab Workflows
Explore how whole slide imaging (WSI) can address the challenges of pathologist shortages by enabling remote diagnostics for intraoperative frozen section consultation and rapid on-site evaluation. Learn about the technology and solutions that facilitate remote support for these critical procedures.
Webinar: Using Telepathology to Advance Patient Care and Lab Workflows
Webinar: Easing Your Transformation to a Digital Workflow
Discover how to overcome obstacles by focusing on the right pre-analytical and diagnostic workflow steps.
Easing Your Transformation to a Digital Workflow