UserName
Viewing profile as user:
Polymedco Products and Resources
Polymedco banner
Delivering speed, accuracy, and flexibility, the PATHFAST Analyzer provides healthcare professionals with lab quality results directly at the point of care. Compact and user-friendly, PATHFAST delivers timely and dependable results to inform clinical decisions, enhancing both patient outcomes and operational efficiency.
Key Benefits:  
  • Lab quality accuracy with Chemiluminescence and Magtration technologies
  • Run up to six simultaneous assays in under 17 minutes
  • Wide biomarker menu: high-sensitivity Troponin I, NT-proBNP, D-Dimer, Myoglobin, CK-MB and hsCRP
  • Use reagents directly from refrigeration with results unaffected by hemolysis
  • Touch-screen controls and LIS integration offer efficient connectivity in emergency departments, STAT labs, core labs, and urgent care facilities
  • Simplify lab operations and reduce training needs
Explore More
Unlocking Rapid Diagnosis: The Power of POC hsTnl, NO-proBNP, and D-Dimer in Emergency Departments
In this engaging session, Dr. Sean-Xavier Neath dives into how POC biomarkers can help rapidly differentiate between life-threatening conditions, streamline ED workflows, and improve patient outcomes.
Watch the PATHFAST Webinar
Accelerate Cardiac Care Decisions
The PATHFAST Analyzer delivers lab quality results at the point of care in under 17 minutes, helping streamline ED workflows and improve patient outcomes by reducing delays in diagnosis.
Cardiac Care Brochure
Efficiency and Accuracy at the Point of Care
The PATHFAST Analyzer provides fast, accurate POC results with intuitive operations, LIS integration, and the flexibility to run up to six assays simultaneously.
Point of Care Brochure
Benefits of NT-proBNP testing on PATHFAST for patients with heart failure treated with ENTRESTO
The FDA-cleared PATHFAST high-sensitivity Troponin I (hs-cTnI-II) assay supports rapid myocardial infraction diagnosis, offering assistance for timely clinical decisions in emergency and acute care settings.
Benefits of NT-proBNP Brochure
Types of Tests
Explore reagents for use with the PATHFAST Analyzer.
Immunoassay Reagents
Coagulation Reagent
Accessories
Browse these accessories for use with the PATHFAST Analyzer.
Request for More Information
Complete this short form to have a representative contact you with more information on the PATHFAST Analyzer and other solutions for your lab and testing.
Required Field *
Required Field *
Country
State/Province