UserName
Viewing profile as user:
Microbiologics KWIK-STIK QC Devices
Microbiologics KWIK-STIK QC Devices
Each Microbiologics™ KWIK-STIK™ QC Device includes everything you need to grow reference cultures: a qualitative lyophilized microorganism pellet, an ampoule of hydrating fluid, and an inoculating swab. This inclusive design can simplify QC testing for lab techs and reduce errors.
KWIK-STIK products are available in packs of two or six, offer over 900 organisms, and can be used for many types of microbiological control testing.
NOTE: Please sign in to your fishersci.com account to view all of the products available for purchase.
Register for a fishersci.com account.