Cryogenic Storage Vials
Small, capped vials designed to withstand ultra-low temperatures; various capacities; may be sterile, non-sterile, and/or autoclavable; includes internally and externally threaded vials; ideal for sample storage in vapor state liquid nitrogen.
Cryogenic storage in the laboratory is used to preserve samples with liquid nitrogen, a method that requires special products that can tolerate low temperatures without compromising the samples. The U.S. National Institute of Standards and Technology defines cryogenics as related to temperatures below −180°C (−292°F).
Cryogenic storage vials, cryogenic storage tubes, and cell culture cryogenic tubes are designed to safely store samples, and may also be used at general cold-storage temperatures.
Cryogenic vials and tubes have some common characteristics. They are made from non-cytotoxic and non-mutagenic plastic resins and are pyrogen-free. They are sold individually or in bulk packaging and in uncapped or non-sterile varieties for less demanding uses. Sterile tubes meet a SAL of 10-6.
Most cryogenic tubes or vials have screw-style closures that ensure the caps stay on in spite of temperature and air pressure changes. Cryo tubes or vials are available with either internal or external threads. Because the caps fit over the outsides of the tubes, external threads help maintain the sterility of samples.
Some cryogenic storage tubes have round bottoms and others are skirted so they can stand alone. Vials may be sized for specific applications, such as those that need to fit inside long tubes or canes during storage.
In addition, many cryogenic storage tubes come with a writing area on the side of the tube for labeling. Color-coding markers and caps also exist to help organize samples by type, researcher, etc.
Special cryogenic storage canisters are used to contain the liquid nitrogen needed for cryogenic storage.
- (1)
- (12)
- (15)
- (15)
- (16)
- (414)
- (6)
- (8)
- (37)
- (449)
- (23)
- (49)
- (10)
- (143)
- (19)
- (7)
- (33)
- (2)
- (198)
- (77)
- (1)
- (2)
- (10)
- (232)
- (9)
- (14)
- (1)
Filtered Search Results
DWK Life Sciences Wheaton™ CryoELITE™ Cryogenic Storage Vials, Externally Threaded
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
CryoELITE Vial with Loctagon skirt provides stability in freestanding position, and locks into the CryoELITE Benchmate Rack, making it easy to open and close the vial with one hand.
| Thread Style | External |
|---|---|
| Product Type | Cryogenic Vial |
| Dnase Free | Yes |
| Diameter (Metric) Outer | 12 mm |
| Color | Clear |
| Diameter (English) Outer | 0.47 in. |
| RNase Free | Yes |
| Material | Polypropylene |
| Closure Type | Screw Cap |
| Closure Material | Polypropylene |
| Base Style | Freestanding |
DWK Life Sciences Wheaton™ CryoELITE™ Cryogenic Vials with Freestanding Bottom
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Large vial footprint allows use of linear barcode or human readable label
Corning™ Polypropylene Cryogenic Vials
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Manufactured from polypropylene for stability in extreme temperature conditions. Corning™ Externally Threaded Cryogenic Vials provide a safe and secure vessel for ultra-low temperature storage of valuable cell cultures and biological samples.
| Dnase Free | Yes |
|---|---|
| Color | Natural |
| No. per Case | 500 |
| RNase Free | Yes |
| Material | Polypropylene |
| Closure Material | Polypropylene |
| Sterility | Sterile |
| Thread Style | External |
| Includes | Silicone washer for tight seal, 50-position polyfoam rack with each case for easy carrying |
| Marking Spot | Yes |
| Diameter (Metric) Outer | 12.7 mm |
| Graduated | Yes |
| Certifications/Compliance | Nonpyrogenic, RNase/DNase-free |
| No. per Pack | 50 |
| Closure Color | Orange |
| Temperature Range (Metric) | -196°C |
Fisherbrand™ Externally and Internally Threaded Cryogenic Storage Vials
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Choice of externally threaded vials with HDPE closures for aseptic technique, or internally threaded vials featuring polypropylene closures with silicone gasket for tight seal
| Material | Polypropylene |
|---|---|
| Autoclavable | Not Autoclavable |
| Closure Type | Screw Cap |
| For Use With (Application) | For storage of biological materials in mechanical freezers or in the vapor phase of liquid Nitrogen |
| Sterility | Sterile |
Thermo Scientific™ Storage Tube Protection
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Give your cryogenic storage tubes and vials extra protection, such as when shipping or storing in liquid or vapor phase nitrogen with these storage accessories.
Thermo Scientific™ Cryo Vial Closure Color Coders
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
| Product Type | Vial Closure |
|---|
DWK Life Sciences Wheaton™ CryoELITE™ Tissue Vials: 5mL, White Cap, Sterile
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Secure, reliable store container for tissue sections
Corning™ CoolCell™ 2mL Filler Vials for CoolCell™ LX and FTS30 Containers
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Suitable for repeated use. Corning™ CoolCell™ 2mL Filler Vials ensure cell freezing rate consistency and uniform results when using CoolCell containers.
Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
| Dnase Free | Yes |
|---|---|
| RNase Free | Yes |
| Material | Polypropylene |
| Product Line | Nunc |
Argos Technologies™ PolarSafe™ Cryogenic Vials
Innovactive screw cap eliminates need for silicone o-rings
| Pre-slit | No |
|---|---|
| Dnase Free | Yes |
| Color | Clear |
| Quantity | 50/Cs. |
| RNase Free | Yes |
| Material | Polyethylene |
| Closure Material | Polyethylene, Thermoplastic Elastomer |
| Sterility | Sterile |
| Thread Style | External |
| Preservative | None |
| Product Type | Cryogenic Vial |
| Marking Spot | Yes |
| Barcode | Serial/consecutive Barcode128 printed on each tube |
| Diameter (Metric) Outer | 12.5 mm |
| Certifications/Compliance | FDA, GMP and ISO quality standards (ISO13485:2004, ISO14644 and 14698) |
| Autoclavable | Autoclavable |
| Closure Type | Screw Cap |
| Temperature Range (Metric) | -196°C to +121°C |
Corning™ Internally Threaded Cryogenic Vials with Color Caps
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Used for ultra-low temperature storage of valuable cell cultures, biological solutions and aqueous solutions
| Dnase Free | Yes |
|---|---|
| Color | Natural |
| No. per Case | 500 |
| Volume (Metric) | 2 mL |
| RNase Free | Yes |
| Material | Polypropylene |
| For Use With (Application) | For ultra low temperature storage of valuable cell cultures, biological solutions and aqueous solutions |
| Closure Material | Polypropylene |
| Sterility | Sterile |
| Thread Style | Internal |
| Includes | Silicone washer, inserts |
| Marking Spot | Yes |
| Temperature Range (English) | -320.8°F |
| Style | Round |
| Graduated | Yes |
| No. per Pack | 50 |
| Temperature Range (Metric) | -196°C |
| Base Style | Round (Self-standing) |
Corning™ Internally Threaded Cryogenic Vials
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Manufactured from polypropylene for stability in extreme temperature conditions. Corning™ Internally Threaded Cryogenic Vials provide a safe and secure vessel for ultra-low temperature storage of valuable cell cultures and biological samples.
| Thread Style | Internal |
|---|---|
| Dnase Free | Yes |
| Marking Spot | Yes |
| Color | Natural |
| RNase Free | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Certifications/Compliance | Nonpyrogenic, RNase/DNase-free |
| No. per Pack | 50 |
| Closure Color | Orange |
| Temperature Range (Metric) | -196°C |
| Closure Material | Polypropylene |
| Sterility | Sterile |
Globe Scientific Cryoclear™ Internal Thread Cryogenic Vials
Globe Scientific Cryoclear™ Internal Thread Cryogenic Vials are designed for cryogenic storage and transport of biological specimens at temperature as low as -196°C.
| Thread Style | Internal |
|---|---|
| Diameter (Metric) Outer | 12.5 mm |
| Color | Natural |
| No. per Case | 500 |
| Graduated | Yes |
| Material | Medical Grade Polypropylene |
| Certifications/Compliance | Lot Certified: Rnase, Dnase, Pyrogen, Himan DNA and ATP Free, Non-Cytotoxic, Non-Mutagenic, Heavy Me |
| Autoclavable | Autoclavable |
| Sterility | Sterile |
Globe Scientific Cryoclear™ External Thread Cryogenic Vials
Globe Scientific Cryoclear™ External Thread Cryogenic Vials are designed for cryogenic storage and transport of biological specimens at temperature as low as -196°C.
| Thread Style | External |
|---|---|
| Diameter (Metric) Outer | 12.5 mm |
| Color | Natural |
| No. per Case | 500 |
| Graduated | Yes |
| Material | Medical Grade Polypropylene |
| Certifications/Compliance | Lot Certified: Rnase, Dnase, Pyrogen, Himan DNA and ATP Free, Non-Cytotoxic, Non-Mutagenic, Heavy Me |
| Autoclavable | Autoclavable |
| Sterility | Sterile |
Corning™ Bar-coded Cryogenic Vials
For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.
| Volume (Metric) | 2 mL |
|---|---|
| For Use With (Application) | Cell biology, cell banking, compound cryopreservation |