Choosing the Right Filtration Products
Using the correct laboratory filter for processing liquids is an important decision, but it does not have to be difficult or complicated. Filter membranes are available in a variety of materials, pore sizes and other specifications, but you can narrow the selection by asking just a few questions.
Why are you filtering?
There are many reasons why a filtration step may be part of your laboratory protocol. Most often, filtration is performed to remove something from the liquid, a function that is dependent on the membrane pore size.
- To remove Mycoplasma from a solution, use a 0.1μm pore filter
 - Routine laboratory sterilization of media, buffers, biological fluids and gases requires a 0.2 or 0.22μm pore filter membrane
 - Clarification or pre-filtration of solutions and solvents can be accomplished with a 0.45μm pore size
 - Glass fiber filters can be used for water testing or as pre-filters (to improve subsequent filter performance)
- Glass fiber filters can be used for water testing or as pre-filters (to improve subsequent filter performance)
What are you filtering?
The chemical or biological characteristics of the liquid will also affect the choice of filters.
- Some membranes contain wetting agents or other non-toxic substances that can interfere with results
 - The membrane material may bind proteins, nucleotides or other macromolecules that can clog the filter, remove important constituents or otherwise compromise the sample
 - Not all filter membranes are compatible with all chemicals, so confirm that the filter that you choose will work with your solvent or solution
 - The temperature and length of exposure to the chemical may also affect the membrane’s integrity
How are you filtering?
The filter choice will also depend on the volume of liquid to be filtered and the method or device that is being used. Filter membranes are available in a variety of formats:
- Flat or disc filters
 - Syringe filters
 - Filter/storage devices
 - Vacuum filters
 - Bottle-top filters
 - Centrifuge or microfuge tube filters
If the membrane is part of a filtration device, confirm that the housing material is also compatible with the liquid being filtered.
| Membrane Material | Physical Characteristics | Chemical Compatibility | Biological Compatibility | Applications | 
|---|---|---|---|---|
| Cellulose Nitrate | 
  | 
  | 
  | 
  | 
| Nylon (Polyamide) | 
  | 
  | 
  | 
  | 
| PES | 
  | 
  | 
  | 
  | 
| Polypropylene | 
  | 
  | - | 
  | 
| PTFE (laminated to PP or PE) | 
  | 
  | - | 
  | 
| PVDF | 
  | 
  | 
  | - | 
| Cellulose Acetate | 
  | 
  | 
  | 
  | 
| Glass Microfiber | 
  | 
  | - | 
  |