Finding the Right HPLC Solvents
HPLC
HPLC is one of the most common analytical procedures performed in labs today, and it’s used to measure a variety of substances, including:
- Explosives, PCBs and VOCs
- Petrochemicals, lubricants and hydrocarbons
- Vitamins and plant extracts
- Sugars, nutritional substances and additives
- Drug impurities, antihistamines, drugs of abuse and NSAIDs
- Fungicides, herbicides and pesticides
Chemical Solvents
Chemical Solvents are a high-volume and critical component of successful HPLC assays, and HPLC-grade chemicals will deliver the best results. The American Chemical Society (ACS) sets standards for various grades of chemicals, and most HPLC-grade chemicals have these characteristics in common:
- Meet ACS specifications
- Submicron filtered
- Purified to contain low contaminants and low residues
- Application-specific and function tested
- Consistent from lot to lot
- Low backgrounds; free of extraneous peaks
- Low UV absorbance in critical ranges
- Fluorescence tested for trace impurities
- Function tested for assay, water and minimal residue after evaporation
The Options
Use standard and affordable HPLC-grade solvents for routine, high-volume and well-characterized applications (QC for pharmaceutical, food and beverage testing). More specific applications may require UHPLC or LC-MS grade chemicals to meet performance requirements. Advantages to using the most appropriate grade of solvents for your application (that can offset the additional cost of a higher purity product):
- Reduces rework
- Offers greater reproducibility in your results
- Facilitates methods transfer
- Improves the sensitivity of instruments
- Reduces erroneous peaks and maximizes peak resolution
- Can extend column life
- Reduces downtime
Your Best Choice
Choose a product line created for a given application or testing methodology.
Pick a manufacturer with the necessary expertise in purification, packaging and analytical testing to produce multiple grades of chemicals with consistent quality at each grade.
Visit fishersci.com/chromatography or fishersci.ca/chromatography for more information about HPLC solvent selection.