UserName
Viewing profile as user:
Gibco Process Liquids Preparation Services
Gibco Bioprocessing Products
Ready-to-Use Buffers and Process Liquids
Gibco™ buffers and process liquids are available in multiple sizes and concentrations to help accelerate the manufacturing process at any scale. Choose from established formulations and packaging options, including bioprocess containers (BPCs), that come with quality documentation to help facilitate efficient and compliant bioprocessing.
  • Meet workflow needs for all stages of biologic development and manufacturing
  • Streamline in-house processes related to formulation, hydration, testing, and validation
  • Mitigate space required for buffer preparation and storage
  • Ease the time requirements dedicated to in-house buffer preparation
Featured Resources
Gibco Water for Injection for Cell Culture Spec Sheet
Find out more about Gibco™ Water for Injection (WFI) for Cell Culture. The pure, cell culture-grade water is used in the production of process liquids, buffers, supplements, media, and cleaning solutions. It is filtered, non-pyrogenic, and available in scalable volumes and bioprocess container configurations to meet your unique manufacturing needs.
Gibco Water for Injection for Cell Culture Spec Sheet
Gibco Sodium Hydroxide Spec Sheet
Discover Gibco™ Sodium Hydroxide (NaOH) solutions, which provide a range of options for security of supply and dual sourcing. Convenient and cost-effective, they come premixed and prepacked in your choice of bioprocessing container and size to meet your scheduling needs and help accelerate your manufacturing.
Gibco Sodium Hydroxide Spec Sheet
Gibco Sodium Chloride Spec Sheet
Explore the benefits of Gibco™ Sodium Chloride (NaCl) solutions, which are sterile filtered, packaged in bioprocess containers, and ready to use. Sourcing NaCl solutions can help you prepare buffers more efficiently so you can focus on additional essential steps in your biopharmaceutical processes.
Gibco Sodium Chloride Spec Sheet
Gibco 20% Ethanol Spec Sheet
Learn more about Gibco™ 20% Ethanol (EtOH), which is sterile filtered, packaged in bioprocess containers, and stocked to be available when you need it. Sourcing Gibco 20% EtOH can streamline your workload for equipment sanitization and column storage, allowing you to focus on the key steps in your downstream processes.
Gibco 20% Ethanol Spec Sheet
About Gibco
As part of Thermo Fisher Scientific, Gibco products and services help empower your work. Choose Gibco media, sera, supplements, buffers, cells, and bioproduction products for their dependability and consistency. Rely on our technical, scientific, and regulatory professionals to help keep your cell therapy, bioproduction, and other research and manufacturing efforts moving forward.