SP Bel-Art Sterileware Product Selection Guide
SP Bel-Art Sterileware Sampling Solutions
Support critical material workflows with a comprehensive selection of scoops, spoons, spatulas, and liquid samplers designed for dependable performance. Manufactured from FDA- and EU-conforming materials for food, pharmaceutical, and cosmetic applications, each SP Bel-Art Sterileware™ tool is lot-stamped to help ensure traceability. With more than 90% of products made in the U.S., the collection offers both trusted quality and reliable availability.
Featured Products
Product Comparison
| Product | Used with | Sample Type | Capacity | FDA Conforming |
|---|---|---|---|---|
| Pharma Scoops | Drums, Pails | Solids | 60–2,500 mL | Yes |
| Volumetric Sampling Spoons | Bottles | Solids, Creams | 1–20 mL | Yes |
| Powder Thieves | Paper Sacks, Drums | Solids | 1–20 mL | Yes |
| Upright Handle Dippers | Tanks, Drums | Liquids, Creams | 50 mL | Yes |
Browse the SP BelArt Catalog
Discover the range of SP BelArt Sterileware Products.
Complementary Products
Catalog Number 16241210
Catalog Number S43257
Catalog Number 22111526
Catalog Number 11865109