UserName
Viewing profile as user:
BD Rhapsody Cell Analysis Products
BD Rhapsody Cell Analysis Products
Targeted RNA-Seq and Multiplexing Technologies
The BD Rhapsody™ portfolio of products helps you answer biological questions using a sensitive, targeted approach to single-cell analysis. Researchers can interrogate gene expression from hundreds to tens of thousands of single cells from one or multiple samples simultaneously.
  • Portable and affordable system for single-cell studies
  • Captures sensitive and fragile cell types to generate unbiased data
  • Enables multiomics studies
  • No power cords or complicated software/training
BD Single-Cell Multiplexing Kit
With this kit, instead of running one sample per single-cell capture, you can pool up to 12 samples to minimize batch effects and reduce time to discovery and experimental costs.
BD Rhapsody Single-Cell Analysis System
This system enables the high-throughput capture of multiomic information from single cells using a simple cartridge workflow and a multitier barcoding system. When used along with the BD Rhapsody Scanner, you can review quality control measures at different stages of the workflow, such as cell capture rate and cell multiplet rate, to make informed decisions or troubleshoot experiments, if necessary, before expensive sequencing.
BD Rhapsody HT Xpress System
This decentralized, portable, easy-to-use option enables single-cell capture, bead-based mRNA isolation, and bead retrieval right at your bench without having to move to a core lab. It generates reliable, unbiased data and is compatible with the BD Rhapsody Scanner for visual workflow quality control.
BD Rhapsody Targeted Single-Cell RNA-Seq Panels
Targeted gene expression assays are an efficient way to screen hundreds of genes at once, which provide a cost-effective, more sensitive detection of low abundant transcripts than WTA assays. With this targeted approach you can remove high expressers to eliminate excessive, costly sequencing reads.
  • Immune Response Panel (Human)
  • Immune Response Panel (Mouse)
  • T-Cell Panel
  • Onco-BC (breast cancer) Panel
  • Custom panel and supplementary panels
Reach Out to a Representative
Fill out the form and a Fisher Healthcare representative will contact you to answer any questions about 3DHISTECH Pannoramic digital slide scanners.
Required Field*
State
Alaska
Alabama
Arkansas
Arizona
California
Colorado
Connecticut
D.C.
Florida
Georgia
Guam
Hawaii
Iowa
Idaho
Illinois
Indiana
Kansas
Kentucky
Louisiana
Massachusetts
Maryland
Maine
Michigan
Minnesota
Missouri
Mississippi
Montana
North Carolina
North Dakota
Nebraska
New Hampshire
New Jersey
New Mexico
Nevada
New York
Ohio
Oklahoma
Oregon
Pennsylvania
Puerto Rico
Palau
Rhode Island
South Carolina
South Dakota
Tennessee
Texas
Utah
Virginia
Virgin Islands
Vermont
Washington
Wisconsin
West Virginia
Wyoming
Military Americas
Military Europe/ME/Canada
Military Pacific
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Thermo Fisher Scientific Logo
Featured Products
Catalog Number BDB633701
Catalog Number BDB666625
Featured Resources
 BD Biosciences logo
Innovations and Advancements
BD Biosciences began in the 1970s at Stanford University where Dr. Leonard Herzenberg invented fluorescence-activated cell sorter (FACS) technology. Since then, they’ve been at the forefront of single-cell analysis.
With a dedication and passion for research and development, BD Biosciences is taking a closer look at what we know about the building blocks of human life and blazing new trails in immunology.
BD Biosciences is committed to being the scientific partner of choice for your laboratory, pushing the boundaries of what’s possible at the cellular level and enabling change through informatics, genomics, proteomics, transcriptomics, and high-resolution imaging.
By setting the stage for the next decade of medical research, BD Biosciences is empowering scientists around the world to unlock the next generation of treatments and improve lives.