Learn More
BD Rhapsody™ Scanner
Catalog No. BDB633701 Shop All Waters Corporation Products
Change view
Click to view available options
Product Line:
BD Rhapsody
Catalog No. Product Line
BDB633701 BD Rhapsody
1 options
This item is not returnable. View return policy
Catalog No. BDB633701 Supplier BD Supplier No. 633701
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The BD Rhapsody™ Scanner is designed to visualize all steps in the single-cell capture workflow and provide detailed analysis metrics at every step, enabling the user to make key decisions throughout the workflow.
  • Designed to visualize all steps in the single-cell capture workflow and provide detailed analysis metrics at every step, enabling the user to make key decisions throughout the workflow
  • Intended for single-cell mRNA capture upstream of library preparation
  • Provides automated QC and cell capture information during the workflow
  • Two custom pipettes are required for use with the Express instrument
  • Does not include reagent
TRUSTED_SUSTAINABILITY
Specifications
Product Line BD Rhapsody