Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AAACAUUCGCGGUGCACUUCUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-543 | MIMAT0004954 | |
| Mouse | mmu-miR-543-3p | MIMAT0003168 | |
| Bovine | bta-miR-543 | MIMAT0009344 | |
| Callithrix jacchus | cja-miR-543 | MIMAT0039339 | |
| Dasypus novemcinctus | dno-miR-543-3p | MIMAT0047972 | |
| Dog | cfa-miR-543 | MIMAT0006720 | |
| View More Rows | |||
| Guinea pig | cpo-miR-543-3p | MIMAT0047344 | |
| Horse | eca-miR-543 | MIMAT0013169 | |
| Pan troglodytes | ptr-miR-543 | MIMAT0008215 | |
| Pongo pygmaeus | ppy-miR-543 | MIMAT0015993 | |
| Pteropus alecto | pal-miR-543-3p | MIMAT0040219 | |
| Rhesus monkey | mml-miR-543-3p | MIMAT0012787 | |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool B v3.0
Related Products
miRBase Alias:
Mouse: mmu-miR-543 (v17)
Macaca mulatta: mml-miR-543 (v19)
Macaca mulatta: mml-miR-543 (v19)

Genomic Map