Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AAACAUUCGCGGUGCACUUCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-543 | MIMAT0004954 |
| Mouse | mmu-miR-543-3p | MIMAT0003168 |
| Bovine | bta-miR-543 | MIMAT0009344 |
| Callithrix jacchus | cja-miR-543 | MIMAT0039339 |
| Dasypus novemcinctus | dno-miR-543-3p | MIMAT0047972 |
| Dog | cfa-miR-543 | MIMAT0006720 |
| View More Rows | ||
| Guinea pig | cpo-miR-543-3p | MIMAT0047344 |
| Horse | eca-miR-543 | MIMAT0013169 |
| Pan troglodytes | ptr-miR-543 | MIMAT0008215 |
| Pongo pygmaeus | ppy-miR-543 | MIMAT0015993 |
| Pteropus alecto | pal-miR-543-3p | MIMAT0040219 |
| Rhesus monkey | mml-miR-543-3p | MIMAT0012787 |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-543 (v17)
Macaca mulatta: mml-miR-543 (v19)
Macaca mulatta: mml-miR-543 (v19)

Genomic Map