Assay Details
Amplicon Length: | 65 |
Chromosome Location: | Chr. 12 - 109717486 - 109717486 [-] on 109717486 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-543-5p | MIMAT0017207 |
| Mouse | mmu-miR-543-3p | MIMAT0003168 |
Stem-loop Details
Mouse

Genomic Map