Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | UAAGUGCUUCCAUGUUUUAGUAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302b-3p | MIMAT0000715 |
| Mouse | mmu-miR-302b-3p | MIMAT0003374 |
| Anolis carolinensis | aca-miR-302b | MIMAT0021919 |
| Bovine | bta-miR-302b | MIMAT0009280 |
| Chicken | gga-miR-302b-3p | MIMAT0003357 |
| Chrysemys picta | cpi-miR-302b-3p | MIMAT0037861 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-302b-3p | MIMAT0047794 |
| Guinea pig | cpo-miR-302b-3p | MIMAT0047149 |
| Horse | eca-miR-302b | MIMAT0012912 |
| Pan troglodytes | ptr-miR-302b | MIMAT0008087 |
| Pongo pygmaeus | ppy-miR-302b | MIMAT0015817 |
| Rhesus monkey | mml-miR-302b | MIMAT0006260 |
Stem-loop Details
Human
Mouse
Anolis carolinensis
Bovine
Chicken
Chrysemys picta
Dasypus novemcinctus
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map