Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | UGCUUCCUUUCAGAGGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-516a-3p | MIMAT0006778 |
| Human | hsa-miR-516b-3p | MIMAT0002860 |
| Gorilla gorilla | ggo-miR-516a-3p | MIMAT0036442 |
| Pan troglodytes | ptr-miR-516a | MIMAT0008182 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes

Genomic Map