Assay Details
Gene Family ID: | MIPF0000097, mir-338 |
Mature miRNA Sequence: | UCCAGCAUCAGUGAUUUUGUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-338-3p | MIMAT0000763 |
| Mouse | mmu-miR-338-3p | MIMAT0000582 |
| Alligator mississippiensis | ami-miR-338-3p | MIMAT0038269 |
| Astatotilapia burtoni | abu-miR-338 | MIMAT0042116 |
| Cyprinus carpio | ccr-miR-338 | MIMAT0026291 |
| Fugu rubripes | fru-miR-338 | MIMAT0003007 |
| View More Rows | ||
| Gadus morhua | gmo-miR-338-3p | MIMAT0044153 |
| Horse | eca-miR-338-3p | MIMAT0013036 |
| Ictalurus punctatus | ipu-miR-338 | MIMAT0029502 |
| Metriaclima zebra | mze-miR-338 | MIMAT0042360 |
| Neolamprologus brichardi | nbr-miR-338 | MIMAT0042544 |
| Oreochromis niloticus | oni-miR-338 | MIMAT0042724 |
| Pan troglodytes | ptr-miR-338 | MIMAT0008106 |
| Pig | ssc-miR-338 | MIMAT0015713 |
| Pongo pygmaeus | ppy-miR-338-3p | MIMAT0015841 |
| Pundamilia nyererei | pny-miR-338 | MIMAT0042940 |
| Rhesus monkey | mml-miR-338-3p | MIMAT0006278 |
| Salmo salar | ssa-miR-338a-3p | MIMAT0032615 |
| Sarcophilus harrisii | sha-miR-338 | MIMAT0022818 |
| Taeniopygia guttata | tgu-miR-338-3p | MIMAT0014542 |
| Tetraodon nigroviridis | tni-miR-338 | MIMAT0003008 |
| Xenopus tropicalis | xtr-miR-338 | MIMAT0003665 |
| Zebrafish | dre-miR-338 | MIMAT0001873 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-338 |
| Stem-loop Accession # | MI0000814 |
| Stem-loop Sequence | UCUCCAACAAUAUCCUGGUGCUGAGUGAUGACUCAGGCGACUCCAGCAUCAGUGAUUUUGUUGAAGA |
| Chromosome Location | Chr. 17 - 81125883 - 81125949 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000763
miRBase ID: hsa-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: Chr. 17 - 81125883 - 81125949 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-338 |
| Stem-loop Accession # | MI0000619 |
| Stem-loop Sequence | CAACGCUGCACAGGCCGUCCUCCCCAACAAUAUCCUGGUGCUGAGUGGGUGCACAGUGACUCCAGCAUCAGUGAUUUUGUUGAAGAGGGCAGCUGCCA |
| Chromosome Location | Chr. 11 - 120014765 - 120014862 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000582
miRBase ID: mmu-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: Chr. 11 - 120014765 - 120014862 [-] on Build GRCm38 |
| Stem-loop ID | ami-mir-338-1 |
| Stem-loop Accession # | MI0029816 |
| Stem-loop Sequence | CCACUUGCUCCCGCCUCCCCAACAAUAUCCUGGUGCUGAGCGAGUUGCGCACAGAGACUCCAGCAUCAGUGAUUUUGUUGAGGAGGGCGAGCUCUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038269
miRBase ID: ami-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ami-mir-338-2 |
| Stem-loop Accession # | MI0029817 |
| Stem-loop Sequence | UGCAAAUGUUUAUUUCUGGCAACACUAUCCUGAUGCUGUCUGAGUAUGUGGUAAAGCUCCAGCAUCAGUGAUUUUGUUGUUAGUGGUAAAUUCAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038269
miRBase ID: ami-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | abu-mir-338-1 |
| Stem-loop Accession # | MI0033821 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGCCUGAGUAGUUGUCACAAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042116
miRBase ID: abu-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | abu-mir-338-2 |
| Stem-loop Accession # | MI0033822 |
| Stem-loop Sequence | AACAACAUCCUGAUGCUGUCUGGGUAUGUAAAGCAGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042116
miRBase ID: abu-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | abu-mir-338-3 |
| Stem-loop Accession # | MI0033820 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCCUGAGUGCUCCCUAAAGACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042116
miRBase ID: abu-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ccr-mir-338 |
| Stem-loop Accession # | MI0023391 |
| Stem-loop Sequence | UGGGAACAAUAUCCUGAUGCUGAAUGAGUGUGUUUUGUAAAACUCCAGCAUCAGUGAUUUUGUUGCCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026291
miRBase ID: ccr-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | fru-mir-338 |
| Stem-loop Accession # | MI0003335 |
| Stem-loop Sequence | GGAUUCUCCCUGCAACAACAUCCUGGUGCUGCCUGAGUCACUUUCACAAACUCCAGCAUCAGUGAUUUUGUUGCGGGGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003007
miRBase ID: fru-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | gmo-mir-338-1 |
| Stem-loop Accession # | MI0036097 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGAUUGAGUAGUUGUGAUAUAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044153
miRBase ID: gmo-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | gmo-mir-338-2 |
| Stem-loop Accession # | MI0036077 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGCCCGGGUGUGUGCAGCAGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044153
miRBase ID: gmo-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | gmo-mir-338-3 |
| Stem-loop Accession # | MI0036202 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCUUGAGUUCUGCUUCCACACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044153
miRBase ID: gmo-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | eca-mir-338 |
| Stem-loop Accession # | MI0012781 |
| Stem-loop Sequence | CAACAAUAUCCUGGUGCUGAGUGAUGACACACGCAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013036
miRBase ID: eca-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ipu-mir-338-1 |
| Stem-loop Accession # | MI0024620 |
| Stem-loop Sequence | AACAAUCUCCUGAUGCUGCCUGAGUGUUUUUCUUCCACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029502
miRBase ID: ipu-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ipu-mir-338-2 |
| Stem-loop Accession # | MI0024621 |
| Stem-loop Sequence | AACAAUAUCCUGAUGCUGAAUGAGUGUGUUGAAGGAAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029502
miRBase ID: ipu-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ipu-mir-338-3 |
| Stem-loop Accession # | MI0024619 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCCUGAGUACAUCUCACAGACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029502
miRBase ID: ipu-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-338-1 |
| Stem-loop Accession # | MI0034134 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGCCUGAGUAGUUGUCACAAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042360
miRBase ID: mze-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-338-2 |
| Stem-loop Accession # | MI0034136 |
| Stem-loop Sequence | AACAACAUCCUGAUGCUGUCUGGGUGUGUAAAGCAGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042360
miRBase ID: mze-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-338-3 |
| Stem-loop Accession # | MI0034135 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCCUGAGUGCUCCCUAAAGACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042360
miRBase ID: mze-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-338-1 |
| Stem-loop Accession # | MI0034393 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGCCUGAGUAGUUGUCACAAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042544
miRBase ID: nbr-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-338-2 |
| Stem-loop Accession # | MI0034395 |
| Stem-loop Sequence | AACAACAUCCUGAUGCUGUCUGGGUGUGUAAAGCAGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042544
miRBase ID: nbr-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-338-3 |
| Stem-loop Accession # | MI0034394 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCCUGAGUGCUCCCUAAAGACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042544
miRBase ID: nbr-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-338-1 |
| Stem-loop Accession # | MI0034641 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGCCUGAGUAGUUGUCACAAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042724
miRBase ID: oni-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-338-2 |
| Stem-loop Accession # | MI0034647 |
| Stem-loop Sequence | AACAACAUCCUGAUGCUGUUUGGGUGUGUAAAGCAGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042724
miRBase ID: oni-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-338-3 |
| Stem-loop Accession # | MI0034642 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCCUGAGUGCUCCUUAAAGACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042724
miRBase ID: oni-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ptr-mir-338 |
| Stem-loop Accession # | MI0008625 |
| Stem-loop Sequence | UCUCCAACAAUAUCCUGGUGCUGAGUGAUGACUCAGGCGACUCCAGCAUCAGUGAUUUUGUUGAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008106
miRBase ID: ptr-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ssc-mir-338 |
| Stem-loop Accession # | MI0014773 |
| Stem-loop Sequence | CCGUCCUCCCCAACAAUAUCCUGGUGCUGAGUGAUGACACACGCAACUCCAGCAUCAGUGAUUUUGUUGAAGAGGGCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015713
miRBase ID: ssc-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ppy-mir-338 |
| Stem-loop Accession # | MI0014902 |
| Stem-loop Sequence | UCUCCAACAAUAUCCUGGUGCUGAGUGAUGACUCAGGCAACUCCAGCAUCAGUGAUUUUGUUGAAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015841
miRBase ID: ppy-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-338-1 |
| Stem-loop Accession # | MI0034924 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGCCUGAGUAGUUGUCACAAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042940
miRBase ID: pny-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-338-2 |
| Stem-loop Accession # | MI0034930 |
| Stem-loop Sequence | AACAACAUCCUGAUGCUGUCUGGGUGUGUAAAGCAGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042940
miRBase ID: pny-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-338-3 |
| Stem-loop Accession # | MI0034922 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCCUGAGUGCUCCCUAAAGACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042940
miRBase ID: pny-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | mml-mir-338 |
| Stem-loop Accession # | MI0007701 |
| Stem-loop Sequence | UCUCCAACAAUAUCCUGGUGCUGAGUGAUGACUCAGGUGACUCCAGCAUCAGUGAUUUUGUUGAAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006278
miRBase ID: mml-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-338a-1 |
| Stem-loop Accession # | MI0026688 |
| Stem-loop Sequence | AACAAUAUCCUGAUGCUGCAUGAGUGUGUCAAGUGGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032615
miRBase ID: ssa-miR-338a-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-338a-2 |
| Stem-loop Accession # | MI0026687 |
| Stem-loop Sequence | AACAAUAUCCUGAUGCUGCAUGAGUGUGUCAAGUAGAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032615
miRBase ID: ssa-miR-338a-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-338a-3 |
| Stem-loop Accession # | MI0026686 |
| Stem-loop Sequence | AACAAUAUCCUGGUGCUGCCUGAGUGCCUCUUCAUAGACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032615
miRBase ID: ssa-miR-338a-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-338a-4 |
| Stem-loop Accession # | MI0026689 |
| Stem-loop Sequence | AACAACAUCCUGGUGCUGCCUGAGUGACUCUCACCAACUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032615
miRBase ID: ssa-miR-338a-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | sha-mir-338 |
| Stem-loop Accession # | MI0019646 |
| Stem-loop Sequence | CACGAGCGGCCAUGGGGGCAGCCUCUCCCCCACCACCACUGCUCAUCUUGGCUUUCCUCCCCAACAAUAUCCUGAUGCUGAGUGAGCGGCACAUAGAGACUCCAGCAUCAGUGAUUUUGUUGAAGAGGGUGGCUGCCGGCUUCUUCCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022818
miRBase ID: sha-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | tgu-mir-338 |
| Stem-loop Accession # | MI0013756 |
| Stem-loop Sequence | AGCAACACUAUCCUGAUGCUGUCAGAGUAAGUGGUAAAGCUCCAGCAUCAGUGAUUUUGUUGUUAGUGGUAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0014542
miRBase ID: tgu-miR-338-3p Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | tni-mir-338 |
| Stem-loop Accession # | MI0003336 |
| Stem-loop Sequence | GGAUUCUCCCUGCAACAACAUCCUGGUGCUGCCUGAGUCACUUUCACAAACUCCAGCAUCAGUGAUUUUGUUGCGGGGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003008
miRBase ID: tni-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | xtr-mir-338-1 |
| Stem-loop Accession # | MI0004906 |
| Stem-loop Sequence | GUCCCCUGGAACAAUAUCCUGAUGCUGAAUGAGUGGGACAUACAUGCUCCAGCAUCAGUGAUUUUGUUGCAGGCGGCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003665
miRBase ID: xtr-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | xtr-mir-338-2 |
| Stem-loop Accession # | MI0004954 |
| Stem-loop Sequence | CAACAAUAUCCUGAUGCCGUCUGAGUGUGCGGGAAAGCUCCAGCAUCAGUGAUUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003665
miRBase ID: xtr-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | dre-mir-338-1 |
| Stem-loop Accession # | MI0002063 |
| Stem-loop Sequence | GGUUUCUCCCUGCAACAAUCUCCUGAUGCUGCCUGAGUGUUUUUCUUCCACUCCAGCAUCAGUGAUUUUGUUGCCGGAGGUCACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001873
miRBase ID: dre-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
| Stem-loop ID | dre-mir-338-2 |
| Stem-loop Accession # | MI0002064 |
| Stem-loop Sequence | GUGUGUGUGUGUGUUUCUGGUGCCUGCUGAGAACAAUAUCCUGAUGCUGAAUGAGUGUGUUGAAGGAAACUCCAGCAUCAGUGAUUUUGUUGCCAGAGGAGCACUUUGGGCAUCCUGUGUAUAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001873
miRBase ID: dre-miR-338 Mature miRNA Sequence: UCCAGCAUCAGUGAUUUUGUUG Chromosome Location: NA |
More Information
Related Products

Genomic Map