Assay Details
Amplicon Length: | 110 |
Chromosome Location: | Chr. 3 - 29416518 - 29416518 [+] on 29416518 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-551b-5p | MIMAT0017236 |
| Mouse | mmu-miR-551b-3p | MIMAT0003890 |
Stem-loop Details
Mouse

Genomic Map