Assay Details
| Gene Family ID: | MIPF0000360, mir-551 | 
| Mature miRNA Sequence: | GAAAUCAAGCUUGGGUGAGACCU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-551b-5p | MIMAT0017236 | 
| Rat | rno-miR-551b-5p | MIMAT0017335 | 
Stem-loop Details
Mouse
Rat
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :