Assay Details
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GAAAUCAAGCUUGGGUGAGACCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-551b-5p | MIMAT0017236 |
| Rat | rno-miR-551b-5p | MIMAT0017335 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-551b* (v17)
Rat: rno-miR-551b* (v18)
Rat: rno-miR-551b* (v18)

Genomic Map