Assay Details
Gene Family ID: | MIPF0000117, mir-139 |
Mature miRNA Sequence: | UGGAGACGCGGCCCUGUUGGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-139-3p | MIMAT0004662 |
| Rat | rno-miR-139-3p | MIMAT0004735 |
Stem-loop Details
Mouse
Rat

Genomic Map