Assay Details
Amplicon Length: | 56 |
Chromosome Location: | Chr. 7 - 101475169 - 101475169 [+] on 101475169 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-139-5p | MIMAT0000656 |
| Mouse | mmu-miR-139-3p | MIMAT0004662 |
Stem-loop Details
Mouse

Genomic Map