Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. 1 - 166589895 - 166589895 [+] on 166589895 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-139-5p | MIMAT0000845 |
| Rat | rno-miR-139-3p | MIMAT0004735 |
Stem-loop Details
Rat

Genomic Map