Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AAGUUGCCCGCGUGUUUUUCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-543-5p | MIMAT0017207 |
| Rat | rno-miR-543-5p | MIMAT0004787 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-543* (v17)
Rat: rno-miR-543 (v18)
Rat: rno-miR-543 (v18)

Genomic Map