Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 2 - 116867899 - 116867899 [-] on 116867899 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-551b-3p | MIMAT0005596 |
| Rat | rno-miR-551b-5p | MIMAT0017335 |
Stem-loop Details
Rat

Genomic Map