Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | UUUGUUCGUUCGGCUCGCGUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-375-3p | MIMAT0000728 | |
| Mouse | mmu-miR-375-3p | MIMAT0000739 | |
| Rat | rno-miR-375-3p | MIMAT0005307 | |
| Dasypus novemcinctus | dno-miR-375-3p | MIMAT0047849 | |
| Dog | cfa-miR-375 | MIMAT0009871 | |
| Gorilla gorilla | ggo-miR-375 | MIMAT0024189 | |
| View More Rows | |||
| Guinea pig | cpo-miR-375-3p | MIMAT0047208 | |
| Pan troglodytes | ptr-miR-375 | MIMAT0008125 | |
| Pig | ssc-miR-375 | MIMAT0041608 | |
| Pongo pygmaeus | ppy-miR-375 | MIMAT0015863 | |
| Pteropus alecto | pal-miR-375-3p | MIMAT0040121 | |
| Rabbit | ocu-miR-375-3p | MIMAT0048411 | |
| Rhesus monkey | mml-miR-375 | MIMAT0006302 | |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-375 (v21)
Mouse: mmu-miR-375 (v17)
Rat: rno-miR-375 (v18)
Mouse: mmu-miR-375 (v17)
Rat: rno-miR-375 (v18)

Genomic Map