Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | UUUGUUCGUUCGGCUCGCGUGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-375-3p | MIMAT0000728 |
Mouse | mmu-miR-375-3p | MIMAT0000739 |
Rat | rno-miR-375-3p | MIMAT0005307 |
Dasypus novemcinctus | dno-miR-375-3p | MIMAT0047849 |
Dog | cfa-miR-375 | MIMAT0009871 |
Gorilla gorilla | ggo-miR-375 | MIMAT0024189 |
View More Rows | ||
Guinea pig | cpo-miR-375-3p | MIMAT0047208 |
Pan troglodytes | ptr-miR-375 | MIMAT0008125 |
Pig | ssc-miR-375 | MIMAT0041608 |
Pongo pygmaeus | ppy-miR-375 | MIMAT0015863 |
Pteropus alecto | pal-miR-375-3p | MIMAT0040121 |
Rabbit | ocu-miR-375-3p | MIMAT0048411 |
Rhesus monkey | mml-miR-375 | MIMAT0006302 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-375 (v21)
Mouse: mmu-miR-375 (v17)
Rat: rno-miR-375 (v18)
Mouse: mmu-miR-375 (v17)
Rat: rno-miR-375 (v18)
