Assay Details
Amplicon Length: | 57 |
Chromosome Location: | Chr. 2 - 219001345 - 219001345 [-] on 219001345 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-375-5p | MIMAT0037313 |
| Human | hsa-miR-375-3p | MIMAT0000728 |
Stem-loop Details
Human

Genomic Map