Assay Details
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | CAGCAGCAAUUCAUGUUUUGAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-424-5p | MIMAT0001341 | |
| Dasypus novemcinctus | dno-miR-424-5p | MIMAT0047882 | |
| Eptesicus fuscus | efu-miR-424 | MIMAT0035126 | |
| Guinea pig | cpo-miR-424-5p | MIMAT0047243 | |
| Horse | eca-miR-424 | MIMAT0013217 | |
| Nomascus leucogenys | nle-miR-424 | MIMAT0049391 | |
| View More Rows | |||
| Pan paniscus | ppa-miR-424 | MIMAT0049085 | |
| Pan troglodytes | ptr-miR-424 | MIMAT0008143 | |
| Pig | ssc-miR-424-5p | MIMAT0013920 | |
| Pongo pygmaeus | ppy-miR-424 | MIMAT0015884 | |
| Rabbit | ocu-miR-424-5p | MIMAT0048443 | |
| Rhesus monkey | mml-miR-424-5p | MIMAT0006323 | |
| Saimiri boliviensis | sbo-miR-424 | MIMAT0049493 | |
Stem-loop Details
Human
Dasypus novemcinctus
Eptesicus fuscus
Guinea pig
Horse
Nomascus leucogenys
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Saimiri boliviensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-424 (v17)
Enterovirus CA55-1988: eca-miR-322 (v14)
Macaca mulatta: mml-miR-424 (v19)
Staphylococcus schleiferi: ssc-miR-424 (v18)
Enterovirus CA55-1988: eca-miR-322 (v14)
Macaca mulatta: mml-miR-424 (v19)
Staphylococcus schleiferi: ssc-miR-424 (v18)

Genomic Map