Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. X - 134546905 - 134546905 [+] on 134546905 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-424-5p | MIMAT0001341 |
| Human | hsa-miR-424-3p | MIMAT0004749 |
Stem-loop Details
Human

Genomic Map