Assay Details
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | CAGCAGCAAUUCAUGUUUUGAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-424-5p | MIMAT0001341 |
| Dasypus novemcinctus | dno-miR-424-5p | MIMAT0047882 |
| Eptesicus fuscus | efu-miR-424 | MIMAT0035126 |
| Guinea pig | cpo-miR-424-5p | MIMAT0047243 |
| Horse | eca-miR-424 | MIMAT0013217 |
| Nomascus leucogenys | nle-miR-424 | MIMAT0049391 |
| View More Rows | ||
| Pan paniscus | ppa-miR-424 | MIMAT0049085 |
| Pan troglodytes | ptr-miR-424 | MIMAT0008143 |
| Pig | ssc-miR-424-5p | MIMAT0013920 |
| Pongo pygmaeus | ppy-miR-424 | MIMAT0015884 |
| Rabbit | ocu-miR-424-5p | MIMAT0048443 |
| Rhesus monkey | mml-miR-424-5p | MIMAT0006323 |
| Saimiri boliviensis | sbo-miR-424 | MIMAT0049493 |
Stem-loop Details
Human
Dasypus novemcinctus
Eptesicus fuscus
Guinea pig
Horse
Nomascus leucogenys
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Saimiri boliviensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-424 (v17)
Enterovirus CA55-1988: eca-miR-322 (v14)
Macaca mulatta: mml-miR-424 (v19)
Staphylococcus schleiferi: ssc-miR-424 (v18)
Enterovirus CA55-1988: eca-miR-322 (v14)
Macaca mulatta: mml-miR-424 (v19)
Staphylococcus schleiferi: ssc-miR-424 (v18)

Genomic Map