Assay Details
Gene Family ID: | MIPF0000211, mir-432 |
Mature miRNA Sequence: | UCUUGGAGUAGGUCAUUGGGUGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-432-5p | MIMAT0002814 | |
| Bovine | bta-miR-432 | MIMAT0009317 | |
| Callithrix jacchus | cja-miR-432 | MIMAT0049326 | |
| Dasypus novemcinctus | dno-miR-432-5p | MIMAT0047888 | |
| Dog | cfa-miR-432 | MIMAT0009904 | |
| Goat | chi-miR-432-5p | MIMAT0036227 | |
| View More Rows | |||
| Guinea pig | cpo-miR-432-5p | MIMAT0047251 | |
| Horse | eca-miR-432 | MIMAT0013157 | |
| Pan troglodytes | ptr-miR-432 | MIMAT0008146 | |
| Pongo pygmaeus | ppy-miR-432 | MIMAT0015888 | |
| Rabbit | ocu-miR-432-5p | MIMAT0048447 | |
| Rhesus monkey | mml-miR-432-5p | MIMAT0006327 | |
| Sheep | oar-miR-432 | MIMAT0001416 | |
Stem-loop Details
Human
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
Sheep
More Information
Related Products
miRBase Alias:
Human: hsa-miR-432 (v17)
Macaca mulatta: mml-miR-432 (v19)
Macaca mulatta: mml-miR-432 (v19)

Genomic Map