Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 14 - 100884308 - 100884308 [-] on 100884308 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-136-5p | MIMAT0000448 |
| Human | hsa-miR-136-3p | MIMAT0004606 |
| Human | hsa-miR-432-5p | MIMAT0002814 |
| Human | hsa-miR-432-3p | MIMAT0002815 |
Stem-loop Details
Human

Genomic Map