Assay Details
Gene Family ID: | MIPF0000211, mir-432 |
Mature miRNA Sequence: | UCUUGGAGUAGGUCAUUGGGUGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-432-5p | MIMAT0002814 |
| Bovine | bta-miR-432 | MIMAT0009317 |
| Callithrix jacchus | cja-miR-432 | MIMAT0049326 |
| Dasypus novemcinctus | dno-miR-432-5p | MIMAT0047888 |
| Dog | cfa-miR-432 | MIMAT0009904 |
| Goat | chi-miR-432-5p | MIMAT0036227 |
| View More Rows | ||
| Guinea pig | cpo-miR-432-5p | MIMAT0047251 |
| Horse | eca-miR-432 | MIMAT0013157 |
| Pan troglodytes | ptr-miR-432 | MIMAT0008146 |
| Pongo pygmaeus | ppy-miR-432 | MIMAT0015888 |
| Rabbit | ocu-miR-432-5p | MIMAT0048447 |
| Rhesus monkey | mml-miR-432-5p | MIMAT0006327 |
| Sheep | oar-miR-432 | MIMAT0001416 |
Stem-loop Details
Human
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
Sheep

Genomic Map