Assay Details
Gene Family ID: | MIPF0000209, mir-362 |
Mature miRNA Sequence: | AAUCCUUGGAACCUAGGUGUGAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-362-5p | MIMAT0000705 | |
| Bovine | bta-miR-362-5p | MIMAT0009298 | |
| Dog | cfa-miR-362 | MIMAT0009886 | |
| Goat | chi-miR-362-5p | MIMAT0036173 | |
| Horse | eca-miR-362-5p | MIMAT0013210 | |
| Pongo pygmaeus | ppy-miR-362-5p | MIMAT0015850 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-362-5p | MIMAT0006288 | |
| Sheep | oar-miR-362 | MIMAT0030060 | |
Stem-loop Details
Human
Bovine
Dog
Goat
Horse
Pongo pygmaeus
Rhesus monkey
Sheep
More Information
Related Products
miRBase Alias:
Human: hsa-miR-362 (v9.2)
Bos taurus: bta-miR-362 (v13)
Bos taurus: bta-miR-362 (v13)

Genomic Map