Assay Details
Gene Family ID: | MIPF0000209, mir-362 |
Mature miRNA Sequence: | AAUCCUUGGAACCUAGGUGUGAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-362-5p | MIMAT0000705 |
| Bovine | bta-miR-362-5p | MIMAT0009298 |
| Dog | cfa-miR-362 | MIMAT0009886 |
| Goat | chi-miR-362-5p | MIMAT0036173 |
| Horse | eca-miR-362-5p | MIMAT0013210 |
| Pongo pygmaeus | ppy-miR-362-5p | MIMAT0015850 |
| View More Rows | ||
| Rhesus monkey | mml-miR-362-5p | MIMAT0006288 |
| Sheep | oar-miR-362 | MIMAT0030060 |
Stem-loop Details
Human
Bovine
Dog
Goat
Horse
Pongo pygmaeus
Rhesus monkey
Sheep

Genomic Map