Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | CUAGACUGAAGCUCCUUGAGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-151a-3p | MIMAT0000757 | |
| Bovine | bta-miR-151-3p | MIMAT0003524 | |
| Dasypus novemcinctus | dno-miR-151-3p | MIMAT0047678 | |
| Goat | chi-miR-151-3p | MIMAT0035982 | |
| Pan troglodytes | ptr-miR-151 | MIMAT0008045 | |
| Pongo pygmaeus | ppy-miR-151a-3p | MIMAT0015776 | |
| View More Rows | |||
| Rabbit | ocu-miR-151-3p | MIMAT0048266 | |
| Rhesus monkey | mml-miR-151-3p | MIMAT0006213 | |
Stem-loop Details
Human
Bovine
Dasypus novemcinctus
Goat
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-151-3p (v17)
Bos taurus: bta-miR-151 (v18)
Pongo pygmaeus: ppy-miR-151-3p (v18)
Bos taurus: bta-miR-151 (v18)
Pongo pygmaeus: ppy-miR-151-3p (v18)

Genomic Map